Learn More
ATCC ATCC 1B7.11 (TIB-191) Hybridoma - Anti-TNP monoclonal antibody
Supplier:  ATCC TIB191
Encompass
ATCC 1B7.11 (TIB-191) is a mouse hybridoma that produces a monoclonal antibody specific for the trinitrophenyl (TNP) hapten. The antibody forms rosettes with TNP-conjugated sheep red blood cells and is supplied as a frozen hybridoma culture for research use.
  • Produces monoclonal antibody reactive with trinitrophenyl (TNP).
  • Forms rosettes with TNP-conjugated sheep red blood cells.
  • Supplied frozen for recovery and culture.
  • Store in the vapor phase of liquid nitrogen; do not store at -70°C.
  • Derived from animals immunized with TNP-KLH.
  • Tested negative for ectromelia virus (mousepox).
Catalog No. NC2958429