Learn More
Hach Company DPD Total Chlorine Reagent, Test 'N Tube Vials, pk/50
Supplier:  Hach Company 2105645
Encompass
For total chlorine determination by the DPD Method. Test 'N Tube Reagent Vials contain pre-measured DPD reagent. Range: 0.09 - 5.00 mg/L as Cl2. Sample Size: 10 mL Pack of 50 vials.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-5395
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.