Learn More
Hach Company Phenols TNTplus Vial Test (5-150 mg/L), 24 Tests
Supplier:  Hach Company TNT868
Encompass
Phenols TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Phenols by the 4-Aminoantipyrine method.
Catalog No. 50-199-8153