Learn More
Apexbio Technology LLC Tobramycin 32986-56-4 10mM (in 1mL H20)
SDP
Supplier:  Apexbio Technology LLC B18565.1
Encompass_Preferred
Tobramycin (CAS 32986-56-4) is a small-molecule inhibitor targeting the bacterial ribosomal 30S subunit It is designed to interfere with protein synthesis thereby inhibiting bacterial growth Tobramycin exerts its biological activity primarily through specific binding to the 30S subunit preventing translation and subsequent bacterial proliferation In in vitro and in vivo studies Tobramycin demonstrates inhibitory activity with reported IC50 values ranging from 0 2 g/mL to 4 g/mL depending on the bacterial strain and experimental conditions Based on these pharmacological properties Tobramycin holds research potential in the investigation of bacterial infection mechanisms including studies of Pseudomonas aeruginosa and models of bacterial pneumonia as well as in the exploration of bacterial resistance mechanisms
Catalog No. 50-000-89871