Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tobramycin (Nebramycin factor 6) | 32986-56-4 | >99.9% | 1 ML
SDP

Supplier:  Medchemexpress LLC HYB044110MM/1ML

Encompass_Preferred

Tobramycin (Nebramycin Factor 6) is a parenterally administered, broad-spectrum aminoglycoside antibiotic. It is widely used for treating moderate to severe bacterial infections caused by sensitive organisms. It can also be used in pneumonia research related to *Pseudomonas aeruginosa*.

  • Parenterally administered, broad-spectrum aminoglycoside antibiotic
  • Used for treating moderate to severe bacterial infections
  • Utilized in pneumonia research related to *Pseudomonas aeruginosa*
  • Purity of >99.9%
  • Soluble in water and DMSO
  • Target: Aminoglycoside

Catalog No. 50-004-07744


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.