Learn More
Apexbio Technology LLC Tobramycin Sulfate 49842-07-1 50mg
SDP
Supplier:  Apexbio Technology LLC B495850
Encompass_Preferred
Tobramycin Sulfate (49842-07-1) is a broad-spectrum aminoglycoside antibiotic targeting bacterial ribosomal subunits It is designed to disrupt protein synthesis thereby inhibiting bacterial growth Tobramycin Sulfate exerts its biological activity primarily through binding to bacterial ribosomal subunits and interfering with protein synthesis It demonstrates inhibitory activity predominantly against Gram-negative bacteria with reported IC50 values typically ranging from 0 5 to 8 g/mL depending on bacterial species and experimental conditions Based on these pharmacological properties Tobramycin Sulfate holds research potential in assessing susceptibility profiles investigating resistance mechanisms and establishing bacterial infection models in biomedical research
Catalog No. 50-000-88717