Learn More
VEOLIA WTS TOC SYSTEM SUITABILITY
Supplier:  VEOLIA WTS STD 7445201
Encompass
M500 System Suitability SetSystem Suitability Set for M500 TOC analyzers withStandard iOS. Includes one Reagent Water Blank - Rw, one Standard Solution- Rs (500 µg C/L as USP Sucrose), and one System Suitability Solution -Rss (500 µg C/L as USP 1,4-Benzoquinone).1 E
Catalog No. NC2894284