Learn More
Medchemexpress LLC Tocainide hydrochloride | 71395-14-7 | 99.49% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB1798A50MG
Encompass_Preferred
Tocainide hydrochloride is a sodium channel blocker that blocks sodium channels in the pain-producing foci in nerve membranes. It is a primary amine analog of lidocaine and can be used for the treatment of tinnitus. It is for research use only and not sold to patients.
  • Blocks sodium channels in pain-producing foci in nerve membranes
  • Can be used for the treatment of tinnitus
Catalog No. 50-004-11409