Learn More
Medchemexpress LLC Toceranib phosphate | 874819-74-6 | MFCD25372034 | 98.5% | 494.45 g/mol | C22H28FN4O6P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY10330A5MG
Encompass_Preferred
Toceranib phosphate is an orally active receptor tyrosine kinase inhibitor used in research and preclinical studies. It exhibits antitumor and antiangiogenic activity by inhibiting PDGFR, VEGFR, and Kit, and is commonly applied in cell-based assays and in vivo tumor models.
  • Orally active receptor tyrosine kinase inhibitor.
  • Potent inhibition of PDGFR, VEGFR, and Kit.
  • Applicable to cell-based assays and in vivo tumor studies.
  • Molecular weight approximately 494.45 g/mol.
  • Supplied in small research quantities for laboratory use.
Catalog No. 50-002-46876