Learn More
STA PHARMACEUTICAL US LLC Tocopherol amidite | 100 g | CAS 142386-74-1 | InChIKey USUSEQXJHLFSON-UHFFFAOYSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10041100GM
Tocopherol amidite is a Amidite reagent (Subcategory: Specialty Linker/End Modifier) sold by WuXi TIDES. Offered in 100 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: 142386-74-1
- MDL: No data
- InChIKey: USUSEQXJHLFSON-UHFFFAOYSA-N
- Molecular Weight: 630.938761998
- Molecular Formula: C38H67N2O3P
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29329900
- Country Of Origin: China
- IUPAC: 2-cyanoethyl (2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl) diisopropylphosphoramidite
- SMILES: CC(CCCC(CCCC(CCCC1(CCC2=C(C(C)=C(C(OP(N(C(C)C)C(C)C)OCCC#N)=C2C)C)O1)C)C)C)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.