Learn More
Medchemexpress LLC (±)-α-Tocopherol nicotinate | 51898-34-1 | 99.8% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0757A5G
Encompass_Preferred
(±)-α-Tocopherol nicotinate, vitamin E - nicotinate, is an orally active fat-soluble antioxidant that prevents lipid peroxidation in cell membranes. It is hydrolyzed in the blood to α-tocopherol and niacin and may be used in studies of related vascular diseases.
  • Prevents lipid peroxidation in cell membranes.
  • Hydrolyzed in the blood to α-tocopherol and niacin.
  • May be used in studies of related vascular diseases.
  • Can help slow the progression of microangiopathy in type 2 diabetics by reducing lipid peroxidation stress in the red blood cell membrane, improving blood rheology and red blood cell deformability.
  • Increases the proportion of CD4+CD8-T cells in thymocytes.
Catalog No. 50-004-09375