Learn More
Medchemexpress LLC Toddalolactone | 483-90-9 | MFCD08693573 | 100.0% | 308.33 | C16H20O6 | 10 MG

Supplier: Medchemexpress LLC HYN051810MG
Toddalolactone is a natural coumarin isolated from Toddalia asiatica and supplied for research use. It inhibits recombinant human plasminogen activator inhibitor-1 (PAI-1) with a reported IC50 of 37.31 μM and is provided as a high-purity research standard.
- Purity 99.97%.
- Chemical formula C16H20O6.
- Molecular weight 308.33 g/mol.
- CAS number 483-90-9.
- Biological activity PAI-1 inhibitor; IC50 = 37.31 μM.
- Solubility DMSO 50 mg/mL; water < 0.1 mg/mL.
- Storage powder -20°C (3 years), 4°C (2 years).
- Available pack sizes 1 mg, 5 mg, 10 mg, 20 mg, 50 mg, 100 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.