Learn More
Medchemexpress LLC HY-N0518 5mg Medchemexpress, Toddalolactone CAS:483-90-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN05185MG
Encompass_Preferred
Medchemexpress, HY-N0518 5mg Toddalolactone CAS:483-90-9 Toddalolactone, a main component of Toddalia asiatica, inhibits the activity of recombinant human plasminogen activator inhibitor-1 (PAI-1), with an IC50 value of 37.31 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2797