Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Todralazine (hydrochloride) | 3778-76-5 | 99.9% | 1 ML
SDP

Supplier:  Medchemexpress LLC HYB1001A10MM/1ML

Encompass_Preferred

Todralazine hydrochloride, also known as Ecarazine hydrochloride, is an anti-hypertensive agent. It functions as a β2AR blocker and exhibits antioxidant and free radical scavenging properties. This product is intended for research use only.

  • Functions as a β2AR blocker
  • Exhibits antioxidant activity
  • Has free radical scavenging activity
  • Recommended storage for solid form: -20°C, sealed, away from moisture
  • Recommended storage for in-solvent form: -80°C for 6 months, or -20°C for 1 month, sealed, away from moisture
  • Soluble in DMSO and water for in vitro use
  • Suitable for various in vivo protocols

Catalog No. 50-004-07485


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.