Learn More
Cayman Chemical TodralazInhydrochlorIde 250mg
SDP
Supplier:  Cayman Chemical 20893250 MG
Encompass_Preferred
A β2-adrenergic receptor antagonist with peripheral vasodilator activity; expanded hematopoietic cell population and protected zebrafish from ionizing radiation (5 µM); inhibited histone acetylation; long-term treatment (3 mg/day for 4 months) impaired hepatocyte histone acetylation and liver regeneration
Catalog No. 50-298-3001