Learn More
Enzo Life Sciences TOFA (10mg). CAS: 54857-86-2
SDP
Supplier:  Enzo Life Sciences BMLEI3210010
Encompass_Preferred
Hypolipidemic agent. Potent, reversible and competitive inhibitor of acetyl-coenzyme A carboxylase (Ac-CoA carboxylase; ACC), a key enzyme involved in fatty acid biosynthesis. It blocks apoptosis induced by the fatty acid synthase inhibitor C75 (ALX-270-286) and stimulates neurite outgrowth and neuronal differentiation in rat pheochromocytoma cells. Alternative name: 5-(Tetradecyloxy)-2-furoic acid, RMI-14514. Purity: ≥99% (TLC). Solubility: Soluble in dimethyl formamide, DMSO (10mg/ml), 100% ethanol or methanol. Sparingly soluble in aqueous buffers. Long Term Storage: -20°C.
Catalog No. 50-201-0708