Learn More
Medchemexpress LLC Tofacitinib | 477600-75-2 | 99.9% | 312.37 | 1 G

Supplier: Medchemexpress LLC HY403541G
Tofacitinib is an orally available JAK3/2/1 inhibitor, with IC50s of 1, 20, and 112 nM, respectively. It is intended for research use only as a laboratory chemical and for the manufacture of substances. This product has not been fully validated for medical applications.
- Identified for research use
- Stable under recommended storage conditions
- Keep container tightly sealed in cool, well-ventilated area
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 2 years or -20°C for 1 year
- Ships at room temperature if less than 2 weeks
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.