Learn More
Medchemexpress LLC Tofacitinib | 477600-75-2 | 99.9% | 312.37 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY403545MG
Encompass_Preferred
Tofacitinib is an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively. It is for research use only.
  • Orally available JAK3/2/1 inhibitor
Catalog No. 50-004-01604