Learn More
Medchemexpress LLC Tofacitinib | 477600-75-2 | 99.96% | 312.37 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY40354R
Encompass_Preferred
Tofacitinib (Standard) is the analytical standard of Tofacitinib, intended for research and analytical applications. It is an orally available JAK3/2/1 inhibitor, commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard for Tofacitinib
  • Intended for research and analytical applications
  • Orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-02363