Learn More
Medchemexpress LLC Tofacitinib (citrate) | 540737-29-9 | 99.9% | 504.49 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY40354AR
Encompass_Preferred
Tofacitinib citrate is an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively. It exhibits antibacterial, antifungal, and antiviral activities.
  • Orally available JAK3/2/1 inhibitor.
  • IC50s: JAK3 (1 nM), JAK2 (20 nM), JAK1 (112 nM).
  • Possesses antibacterial, antifungal, and antiviral activities.
Catalog No. 50-004-02212