Learn More
Medchemexpress LLC Tofacitinib citrate | 540737-29-9 | 99.9% | 504.49 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY40354AR5MG
Encompass_Preferred
Tofacitinib citrate (Standard) is an analytical standard primarily used for research and analytical applications. This compound acts as an orally available JAK1/2/3 inhibitor, demonstrating activity against bacterial, fungal, and viral agents.
  • Orally available JAK1/2/3 inhibitor.
  • Exhibits antibacterial, antifungal and antiviral properties.
  • Suitable for qualitative and quantitative research experiments.
  • Utilized in analytical techniques such as HPLC, GC, and MS.
Catalog No. 50-004-04775