Learn More
Medchemexpress LLC Tofacitinib impurity 2 | 95.4% | 436.69 | 250 MG

Supplier: Medchemexpress LLC 250MGHY40349A
Tofacitinib impurity 2 is an impurity of Tofacitinib. It is intended for research use only and not for sale to patients.
- Purity: 95.42%
- Molecular weight: 436.69
- Formula: C28H44N4
- Appearance: Liquid
- Shipping: Room temperature in continental US; may vary elsewhere.
- Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.