Learn More
Medchemexpress LLC Tofacitinib Impurity T | 1675248-19-7 | 99.8% | C16H22N6O2 | 5 MG

Supplier: Medchemexpress LLC HY785585MG
Tofacitinib Impurity T is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only.
- Drug intermediate for synthesis
- For research use only
- Molecular Weight: 330.39
- Appearance: Solid
- Shipping: Room temperature (continental US)
- Storage: -20°C for 3 years (powder), -80°C for 6 months (in solvent)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.