Learn More
Medchemexpress LLC Tofacitinib Impurity T | 1675248-19-7 | 99.8% | C16H22N6O2 | 50 MG

Supplier: Medchemexpress LLC HY7855850MG
Tofacitinib Impurity T is a drug intermediate, also known as 3-((3R,4R)-4-Methyl-3-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)-3-oxopropanamide. This compound is suitable for the synthesis of various active compounds.
- Purity of 99.8%
- Solid appearance
- Ships at room temperature within continental US
- Storage as powder: -20°C for 3 years, 4°C for 2 years
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.