Learn More
Medchemexpress LLC Tofacitinib (Standard) | 477600-75-2 | 99.96% | 312.37 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY40354R50MG
Encompass_Preferred
Tofacitinib (Standard) is the analytical standard of Tofacitinib, intended for research and analytical applications. Tofacitinib is an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively. It is for research use only and not sold to patients.
  • Analytical standard grade.
  • Used as a reference standard for assays.
  • Commonly employed in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-03843