Learn More
Medchemexpress LLC Tofisopam | 22345-47-7 | 99.46% | C22H26N2O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYA016550MG
Encompass_Preferred
Tofisopam is an orally active 2,3-benzodiazepine compound and anxiolytic agent. It inhibits phosphodiesterase (PDE) isoenzyme activity and is used for the study of anxiety. It acts as an anxiolytic without sedative-hypnotic, muscle relaxant, or anticonvulsive effects, and has been shown to improve cognitive performance and synaptic transmission in amnesic rats.
  • Inhibits phosphodiesterase isoenzyme activity
  • Orally active anxiolytic agent
  • Used for the study of anxiety
  • Does not cause sedative-hypnotic, muscle relaxant, or anticonvulsive effects
  • Active in mouse models of negative symptoms of psychosis
  • Improves impaired cognitive performance
  • Enhances attenuated synaptic transmission in the hippocampus
  • Increases proliferation in subgranular zones
  • Improves the decrease in astrocytes in amnesic rats
Catalog No. 50-004-32945