Learn More
Grainger Toilet Bowl Elongated Floor Gravity Tank
Supplier:  Grainger 16T996
Encompass_Preferred
The American Standard Cadet 3 Right Height Elongated Toilet Bowl is a vitreous china toilet bowl with an elongated front. It features a 12" rough-in and a 16-1/2" rim height. This toilet bowl has a 2-3/8" trapway and is designed for use with the Cadet 3 tank. The Cadet 3 bowl meets EPA WaterSense criteria to use 20% less water than the current federal standard of 1.6 gallons per flush. This is an ADA compliant toilet.
Catalog No. 50-276-6643