Learn More
MSC Royce Rolls Toilet Paper Dispenser, One-Roll, Stainless Steel, Standard
Supplier:  MSC 63344089
Encompass
This standard toilet paper dispenser is designed for durability and hygiene, specifically the one-roll option within the Standard TP Series. Constructed from indestructible stainless steel, it is suitable for various environments.
  • Heavy-duty stainless steel construction
  • Long-lasting and resistant to dents, scratches, and rust
  • Non-porous, flat surface for easy disinfection, offering antibacterial properties against various pathogens
  • Durable 20 gauge, type 304 stainless steel frame with a 1 1/8″ diameter tube
  • Easy to install, sanitize, and refill
  • Security options available separately
  • Backed by a 10-year guarantee
  • Made in the USA
Catalog No. 50-283-7812