Learn More
Medchemexpress LLC Tolazamide | 1156-19-0 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB092050MG
Encompass_Preferred
Tolazamide is an orally active sulfonylurea agent that inhibits sulfonylurea receptor 1 (SUR1) linked to the inwardly rectifying potassium channel. It possesses anti-diabetic properties, capable of lowering blood glucose, decreasing insulin dose while maintaining adequate metabolic control, and improving or normalizing hyperglycemia and HbA. This product is intended for research use only and is not sold to patients.
  • Inhibits sulfonylurea receptor 1 (SUR1)
  • Possesses anti-diabetic properties
  • Lowers blood glucose
  • Decreases insulin dose while maintaining adequate metabolic control
  • Improves or normalizes hyperglycemia and HbA
  • For research use only
Catalog No. 50-004-09081