Learn More
TARGETMOL CHEMICALS INC TOLCAPONE 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6708100MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tolcapone (Ro 40-7592) is a catechol-O-methyltransferase inhibitor used in the therapy of Parkinson disease as adjunctive therapy in combination with levodopa and carbidopa. purity: 99%
Catalog No. 50-257-6470