Learn More
Apexbio Technology LLC Tolcapone, 10mg. Cas: 134308-13-7 MFCD: MFCD00866569. COMT inhibitor
SDP
Supplier:  Apexbio Technology LLC A438310
Encompass_Preferred
Tolcapone is a novel, reversible and orally-bioavailable small-molecule catechol-O-methyltransferase (COMT) inhibitor used for as an adjunct to levodopa therapy for the treatment of Parkinsons disease (PD). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4937