Learn More
Medchemexpress LLC Tolcapone | 134308-13-7 | 99.9% | 273.24 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY174065MG
Encompass_Preferred
Tolcapone (Ro 40-7592) is a selective, potent, and orally active COMT inhibitor with an IC50 of 773 nM. It inhibits α-syn and Aβ42 oligomerization and fibrillogenesis, and can induce oxidative stress, causing apoptosis and ROS production in cancer cells. It is used for research related to cancer and neurological diseases, such as Parkinson's disease and neuroblastoma.
  • Inhibits α-synuclein fibrillization.
  • Inhibits α-syn seed-induced polymerization.
  • Inhibits Aβ42 fibrillization.
  • Inhibits Aβ42 seed-induced polymerization.
  • Protects PC12 cells against α-syn and Aβ42-induced toxicity.
  • Induces apoptosis in neuroblastoma cells.
  • Increases reactive oxygen species levels in cells.
  • Decreases the ATP per cell ratio in neuroblastoma cells.
  • Inhibits tumor growth and prolongs survival in xenograft mouse models.
Catalog No. 50-003-85964