Learn More
Medchemexpress LLC Tolcapone-d4 (ro 40-7592-d4) | 1246816-93-2 | 99.1% | 277.27 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17406S11MG
Encompass_Preferred
Tolcapone-d4 (Ro 40-7592-d4) is the deuterium labeled Tolcapone (HY-17406). Tolcapone (Ro 40-7592) is a selective, potent and orally active COMT inhibitor with an IC50 of 773 nM. Tolcapone can inhibit α-syn and Aβ42 oligomerization and fibrillogenesis. Tolcapone can cause oxidative stress and induce cancer cells apoptosis and ROS production. Tolcapone can be used for the researches of cancer and neurological disease, such as Parkinson disease and neuroblastoma.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-84601