Learn More
Cayman Chemical TolebrutInIb 10mg
SDP
Supplier:  Cayman Chemical 3772310 MG
Encompass_Preferred
An irreversible inhibitor of BTK (IC50 = 1 nM); decreases the number of CD69+ B cells in isolated whole mouse blood (IC50 = 200 nM)
Catalog No. 50-291-6275