Learn More
Medchemexpress LLC HY-109192 5mg Medchemexpress, Tolebrutinib CAS:1971920-73-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1091925MG
Encompass_Preferred
Medchemexpress, HY-109192 5mg Tolebrutinib CAS:1971920-73-6 Tolebrutinib (SAR442168) is a potent, selective, orally active and brain-penetrant inhibitor of Bruton tyrosine kinase (BTK), with IC50s of 0.4 and 0.7 nM in Ramos B cells and in HMC microglia cells, respectively. Tolebrutinib exhibits efficacy in central nervous system immunity. Tolebrutinib can be used for the research of multiple sclerosis (MS). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0768