Learn More
Medchemexpress LLC Tolnaftate | 2398-96-1 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0370R50MG
Encompass_Preferred
Tolnaftate (Standard) is an analytical standard of Tolnaftate, a synthetic thiocarbamate used as an anti-fungal agent. This product is intended for research and analytical applications, and works by blocking sterol biosynthesis in fungal cells.
  • Analytical standard for Tolnaftate
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Functions as an anti-fungal agent
  • Inhibits sterol biosynthesis in fungal cells
Catalog No. 50-004-06275