Learn More
Apexbio Technology LLC Tolperisone HCl 3644-61-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B18575.1
Encompass_Preferred
Tolperisone HCl (CAS 3644-61-9) is a small-molecule inhibitor targeting voltage-gated sodium channels It is designed to inhibit sodium ion influx thereby modulating neuronal excitability and suppressing neuromuscular transmission Tolperisone HCl exerts its biological activity primarily through the inhibition of voltage-gated sodium channels interfering with neuronal signal propagation and affecting muscle tone and contraction Based on these pharmacological properties Tolperisone HCl holds research potential in studies of muscle spasticity neuromuscular disorders and the physiological basis of muscle relaxation and neuromuscular modulation
Catalog No. 50-000-90229