Learn More
Medchemexpress LLC Tolperisone (hydrochloride) | 3644-61-9 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB113910MM/1ML
Encompass_Preferred
Tolperisone hydrochloride is a centrally acting muscle relaxant studied for its use in neurological disorders that cause pathological rhabdomyosclerosis (such as pyramidal tract injury, multiple sclerosis, myelopathy, encephalomyelitis), spastic paralysis, and other types of muscle dystonia-related encephalopathy. Additionally, it exhibits antiviral activity.
  • Induces UPR-mediated cancer cell death by targeting LSD1
  • Inhibits LSD1 activity
  • Induces cancer cell cycle arrest and apoptosis
  • Repolarizes M2 macrophages into M1 phenotype
  • Shows antinociceptive effect in neuropathic pain models
  • Inhibits tumor growth in xenograft mouse models
Catalog No. 50-004-11579