Learn More
Medchemexpress LLC Tolrestat | 82964-04-3 | 99.4% | 357.35 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1650010MM/1ML
Encompass_Preferred
Tolrestat is a potent, orally active aldose reductase inhibitor. It is supplied as a 10 mM solution in 1 mL of DMSO, ready for reconstitution.
  • Potent and orally active aldose reductase inhibitor
  • Has an IC50 of 35 nM
  • Supplied as a 10 mM solution in 1 mL of DMSO
  • Normalizes RBC sorbitol levels in diabetic rats in in vivo studies
  • Inhibits tissue AR activity without affecting plasma lipoprotein levels or body weight
Catalog No. 50-003-94674