Learn More
Medchemexpress LLC Tolrestat | 82964-04-3 | 99.4% | 357.35 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16500
Encompass_Preferred
Tolrestat is a potent, orally active aldose reductase inhibitor with an IC50 of 35 nM. It is intended for research use only.
  • Potent, orally active aldose reductase inhibitor.
  • IC50: 35 nM (Aldose Reductase).
  • Causes a reversal to normal RBC sorbitol levels in diabetic rats.
  • Inhibits tissue AR activity without significantly affecting plasma lipoprotein levels, body weight, or general health in mice.
Catalog No. 50-003-94790