Learn More
Medchemexpress LLC Tolrestat | 82964-04-3 | 99.4% | 357.35 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1650050MG
Encompass_Preferred
Tolrestat is a potent, orally active aldose reductase inhibitor with an IC50 of 35 nM.
  • Potent, orally active aldose reductase inhibitor with IC50 of 35 nM.
  • Causes a reversal to normal RBC sorbitol levels in diabetic rats at 1.8 mg/kg per day.
  • Inhibits tissue AR activity without significantly affecting plasma lipoprotein levels or body weight in mice.
Catalog No. 50-003-94258