Learn More
Medchemexpress LLC Toltrazuril | 69004-03-1 | 99.5% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB01751G
Encompass_Preferred
Toltrazuril is an antiprotozoal agent specifically targeting Coccidia parasites. It operates by inducing structural changes in coccidian developmental stages, affecting the endoplasmic reticulum, Golgi apparatus, and peri-nuclear space, leading to disturbed nuclear division.
  • Acts upon Coccidia parasites
  • Induces structural changes in coccidian developmental stages
  • Affects endoplasmic reticulum and Golgi apparatus
  • Causes abnormalities in the peri-nuclear space
  • Disturbs nuclear division
  • Demonstrates high efficacy in fecal oocyst count reduction
Catalog No. 50-004-05991