Learn More
Medchemexpress LLC Toltrazuril | 69004-03-1 | 99.5% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB017510MM/1ML
Encompass_Preferred
Toltrazuril (BAY-i 9142) is a potent antiprotozoal agent specifically targeting Coccidia parasites. It operates by inducing significant changes in the fine structure of coccidian development stages, causing swelling of the endoplasmic reticulum and Golgi apparatus, alongside abnormalities in the peri-nuclear space and disruptions in nuclear division. This mechanism leads to a substantial reduction in oocyst counts, demonstrating high efficacy against coccidial infections.
  • Acts as an effective antiprotozoal agent against Coccidia parasites.
  • Induces structural changes in coccidian development stages.
  • Significantly reduces oocyst counts in treated subjects.
  • Demonstrates high and sustained efficacy over treatment periods.
Catalog No. 50-004-05350