Learn More
Sigma Aldrich Fine Chemicals Biosciences Toluene ACS reagent 99.5
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 17941818LCS
Encompass
Toluene a flammable liquid with a pungent odor is widely employed as organic solvent. It is widely used as a precursor for synthesizing benzene and as a solvent in the paint industry. It has been reported to be a biotoxic solvent (toxic to many microorganisms at 0.1%v/v concentrations). Its anaerobic biodegradation to CO2 by the denitrifying bacterium Thauera arornatica has been reported. It forms a syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.
Catalog No. 50-289-9195