Learn More
Abcam 3,5-Di-O-(p-toluyl)-2-deo x y-D-ribofuranosyl chloride, 1G
Supplier:  Abcam AB2750541G
Encompass_Preferred
MW 388.8 Da. Ribose analog used as an intermediate for synthesis of deoxynucleotides. Reacts with nucleophilic bases to produce beta-D-ribofuranosyl glycosides.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-079