Learn More
Medchemexpress LLC Tolvaptan-d7 | 1246818-18-7 | 455.98 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17000S1MG
Encompass_Preferred
Tolvaptan-d7 is the deuterium labeled Tolvaptan, a selective, competitive arginine vasopressin receptor 2 antagonist with an IC50 of 1.28μM for the inhibition of AVP-induced platelet aggregation. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs due to deuteration.
  • Selective, competitive arginine vasopressin receptor 2 antagonist.
Catalog No. 50-003-90675