Learn More
Enzo Life Sciences Tomatidine . HCl, (25mg), CAS Number: 6192-62-7
SDP
Supplier:  Enzo Life Sciences BMLGR3350025
Encompass_Preferred
Negative control for Cyclopamine. Steroidal alkaloid structurally similar to cyclopamine (Prod. No. ALX-430-159) but does not inhibit the hedgehog (shh) pathway. May be used as a negative control for jervine (Prod. No. ALX-350-378). Formula: C27H45NO2. HCl. MW: 452.1. CAS Number: 6192-62-7. Purity: ≥90% (TLC). Appearance: Off-white solid. Solubility: Soluble in DMSO (2mg/ml), 100% ethanol (2.5mg/ml) or dimethyl formamide (2mg/ml). Slight warming may be required. Use/Stability: Stock solutions are stable for up to 3 months when stored at -20°C. Long Term Storage: -20°C
Catalog No. NC0985032