Learn More
Medchemexpress LLC Tomatidine hydrochloride | 6192-62-7 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN2149A100MG
Encompass_Preferred
Tomatidine hydrochloride is an anti-inflammatory agent that blocks NF-κB and JNK signaling pathways. It also activates autophagy in mammal cells and C. elegans. In vitro, it decreases inducible NO synthase and COX-2 expression by suppressing I-κBα phosphorylation, NF-κB nuclear translocation, and JNK activation.
  • Blocks NF-κB and JNK signaling.
  • Activates autophagy in mammal cells and C. elegans.
  • Decreases inducible NO synthase and COX-2 expression.
  • Suppresses I-κBα phosphorylation.
  • Inhibits NF-κB nuclear translocation.
  • Inhibits JNK activation.
  • Causes dose-dependent inhibition of LPS-induced iNOS expression.
  • Suppresses NF-κB binding activity.
Catalog No. 50-004-21863