Learn More
Medchemexpress LLC HY-N2166 20mg , Tomatine CAS:17406-45-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN216620MG
Encompass_Preferred
HY-N2166 20mg Tomatine CAS: 17406-45-0 Tomatine is a glycoalkaloid, found in the tomato plant (Lycopersicon esculentum Mill.). Tomatine elicits neurotoxicity in RIP1 kinase and caspase-independent manner. Tomatine promotes the upregulation of nuclear apoptosis inducing factor (AIF) in neuroblastoma cells. Tomatine also inhibits 20S proteasome activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1555