Learn More
Medchemexpress LLC Tominersen | 1709886-74-7 | 96.3% | 10MG

Supplier: Medchemexpress LLC HY13257910MG
Tominersen is a second-generation 2′-O-(2-methoxyethyl) antisense oligonucleotide designed to bind huntingtin (HTT) mRNA and reduce huntingtin protein expression. It is supplied as a solid powder for research use with defined purity and storage recommendations.
- Targets huntingtin (HTT) mRNA to lower mutant huntingtin protein levels.
- Second-generation 2′-O-(2-methoxyethyl) antisense chemistry.
- Supplied as a solid powder suitable for research applications.
- Purity 96.3% as reported by supplier.
- Molecular weight 7078 Da.
- Recommended storage: -20°C; in solvent, -80°C for long-term stability.
- Available in small-scale amounts, including 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.